Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl-Histone H4 (Lys5) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07327
Description
Use Anti-acetyl-Histone H4 (Lys5) Antibody (Rabbit Polyclonal Antibody) validated in western blot, DB, immunocytochemistry, ChIP-seq, Chimmunoprecipitation to detect acetyl-Histone H4 (Lys5) also known as H4K5Ac, Histone H4 (acetyl K5).
Specifications
acetyl-Histone H4 (Lys5) | |
Polyclonal | |
Immunodepleted rabbit serum with 0.05% soduim azide and 30% glycerol. | |
Rabbit | |
Purified by immunodepletion | |
Primary | |
Human | |
Serum |
ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Immunocytochemistry, Western Blot | |
Unconjugated | |
HIST1H4A; H4/A; H4FA; HIST1H4B; H4/I; H4FI; HIST1H4C; H4/G; H4FG; HIST1H4D; H4/B; H4FB; HIST1H4E; H4/J; H4FJ; HIST1H4F; H4/C; H4FC; HIST1H4H; H4/H; H4FH; HIST1H4I; H4/M; H4FM; HIST1H4J; H4/E; H4FE; HIST1H4K; H4/D; H4FD; HIST1H4L; H4/K; H4FK; HIST1H4L; H4/K; H4FK; HIST1H4L; H4/K; H4FK; HIST2H4A | |
KLH-conjugated linear peptide corresponding to the N-terminus of Histone H4 acetylated at Lys5. | |
100 μL | |
NP_001029249 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction