Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Histone H4 (Lys5) Rabbit anti-Human, Vertebrates, MilliporeSigma™

Catalog No. 04118MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-118-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-118-MI Supplier MilliporeSigma Supplier No. 04118
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Detect acetyl-Histone H4 (Lys5) using this Anti-acetyl-Histone H4 (Lys5) Antibody, rabbit validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H4 (Lys5)
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation 100μL of Rabbit Monoclonal IgG cell culture supernatant in 0.1% sodium azide.
Gene Accession No. NM_003538.3
Host Species Rabbit
Immunogen Peptide corresponding to Histone H4 containing the sequence [GRG-AcK-GGK] on which Lys5 is acetylated.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) HIST1H4A, H4/J, HIST1H4F, HIST1H4L, H4FN, H4FA, H4FH, HIST1H4H, H4F2, H4FM, H4/A, H4FK, H4FG, HIST2H4, H4FB, H4/K, HIST1H4I, H4/N, HIST1H4B, H4FE, H4/M, H4/a, HIST1H4E, HIST4H4, HIST2H4A, H4FC, H4FI, H4/H, HIST1H4C, H4/G, HIST1H4K, H4FJ, H4FD, H4/I, H4/B,
Target Species Human, Vertebrates
Content And Storage −20°C for 24 months
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.