Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Histone H4 (Lys8) Rabbit anti-Human, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™

Catalog No. 07328MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-328-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-328-MI Supplier MilliporeSigma Supplier No. 07328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-acetyl-Histone H4 (Lys8) Antibody (Rabbit Polyclonal Antibody) validated in Chimmunoprecipitation, immunoprecipitation, western blot, DB, ChIP-seq to detect acetyl-Histone H4 (Lys8) also known as H4K8Ac, Histone H4 (acetyl K8).
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H4 (Lys8)
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Unpurified rabbit polyclonal in serum with 0.05% sodium azide.
Gene Symbols HIST1H4A; H4/J; HIST1H4F; HIST1H4L; H4FN; H4FA; H4FH; HIST1H4H; H4F2; H4FM; H4/A; H4FK; H4FG; HIST2H4; H4FB; H4/K; HIST1H4I; H4/N; HIST1H4B; H4FE; H4/M; H4/a; HIST1H4E; HIST4H4; HIST2H4A; H4FC; H4FI; H4/H; HIST1H4C; H4/G; HIST1H4K; H4FJ; H4FD; H4/I; H4/B; H4/D; H4/C; HIST1H4J; HIST1H4D; H4/E
Host Species Rabbit
Immunogen Peptide corresponding to amino acids 3-13 of yeast Histone H4 (RGKGGAcKGLGKG-C) in which lysine 8 is acetylated.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_175054
Target Species Human, S. cerevisiae, Yeast
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.