Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Histone H4 Rabbit anti-Human, Phycoerythrin, Polyclonal, MilliporeSigma™

Catalog No. FCABS326PE Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCABS326PE 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCABS326PE Supplier MilliporeSigma Supplier No. FCABS326PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Milli-Mark™™ Anti-acetyl-Histone H4 Antibody-PE is a highly specific rabbit polyclonal antibody, that targets Histone H4 and has been tested in Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H4
Applications Flow Cytometry
Classification Polyclonal
Conjugate PE
Formulation Purified rabbit polyclonal IgG conjugated to PE in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Symbols HHF1; HHF2
Host Species Rabbit
Immunogen Tetrahymena histone H4 acetylated on lysines 5, 8, 12, and 16.
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) XP_001016593
Target Species Human
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.