Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl (Lys34) phospho (Ser36) Histone H1 Rabbit anti-Bovine, Human, Polyclonal, MilliporeSigma™

Catalog No. 061358MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-135-8MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-135-8MI Supplier MilliporeSigma Supplier No. 061358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-acetyl (Lys34) phospho (Ser36) Histone H1 Antibody (Rabbit Polyclonal Antibody) validated in western blotting and DB to detect acetyl (Lys34) phospho (Ser36) Histone H1 also known as Histone H1b.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl (Lys34) phospho (Ser36) Histone H1
Applications Dot Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols HIST1H1E; H1F4
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Histone H1 acetylated at Lys34 and phosphorylated at Ser36.
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_005312
Target Species Bovine, Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.