Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl-Lysine Mouse anti-Mouse, Rat, Human, Vertebrates, Clone: 4G12, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05515
Description
Detect acetyl-Lysine with Anti-acetyl-Lysine Antibody, clone 4G12 (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation and western blotting.
Specifications
| acetyl-Lysine | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| Protein G chromatography | |
| Primary | |
| −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| 4G12 | |
| Purified mouse monoclonal IgG in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide, before the addition of glycerol to 30%. | |
| Mixture of chemically acetylated antigens; recognizes numerous proteins acetylated on a lysine, most prominently on histones. | |
| 100 μg | |
| Human, Mouse, Rat, Vertebrates | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction