Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Lysine Rabbit anti-Vertebrates, Polyclonal, MilliporeSigma™

Catalog No. 06933MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-933-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-933-MI Supplier MilliporeSigma Supplier No. 06933
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-acetyl-Lysine Antibody is a high quality Rabbit Polyclonal Antibody for the detection of acetyl-Lysine and has been validated in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Lysine
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit IgG serum containing 0.05% sodium azide and 30% glycerol.
Host Species Rabbit
Immunogen A mixture of chemically acetylated antigens.
Purification Method Antiserum
Quantity 100 μL
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage Stable for 1 year at .
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.