Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl Lysine Rabbit anti-All, Polyclonal, MilliporeSigma™

Catalog No. AB3879MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3879MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3879MI Supplier MilliporeSigma Supplier No. AB3879
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Acetyl Lysine Antibody is a high quality Rabbit Polyclonal Antibody for the detection of Acetyl Lysine and has been validated in ELISA, immunofluoresence, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl Lysine
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with 50% glycerol.
Host Species Rabbit
Immunogen Acetylated KLH.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species All species
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.