Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-p53 (Lys120) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE286MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE286MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE286MI Supplier MilliporeSigma Supplier No. ABE286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-acetyl-p53 (Lys120) Antibody is a highly specific rabbit polyclonal antibody, that targets p53 and has been tested in Dot Blot and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-p53 (Lys120)
Applications Dot Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols TP53; P53
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the AXIN1 interacting domain of human p53 acetylated at Lys120.
Purification Method Affinity Purified
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_000537
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.