Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-p53 Rabbit anti-Vertebrates, Polyclonal, MilliporeSigma™

Catalog No. 06758MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-758-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-758-MI Supplier MilliporeSigma Supplier No. 06758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect acetyl-p53 (Lys373) with Anti-acetyl-p53 Antibody (Lys373, Lys382) (Rabbit Polyclonal Antibody), that has been shown to work in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-p53 (Lys373, Lys382)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antiserum containing 0.05% sodium azide. Frozen solution.
Gene Symbols TP53; P53; TRP53; p53; LFS1
Host Species Rabbit
Immunogen peptide of human p53 corresponding to amino acids 368-389 with a duplication of amino acids 376 and 377 (HLKSKACKGQSTSTSRHKACKLMFKTC) where ACK denotes acetyl-lysine
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000546
Target Species Vertebrates
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.