Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-p53 (Lys373) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 06916MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-916-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-916-MI Supplier MilliporeSigma Supplier No. 06916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-acetyl-p53 (Lys373) Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-p53 (Lys373) also known as Antigen NY-CO-13, Phosphoprotein p53, Tumor suppressor p53, p53 antigen and has been tested in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-p53 (Lys373)
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.02% sodium azide, 0.1mg/ml BSA
Gene Symbols TP53; P53; TRP53; p53; LFS1
Host Species Rabbit
Immunogen peptide corresponding to amino acids 367-379 of human p53 (SHLKSKACKGQSTSR, where ACK denotes acetyl-lysine).
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000546
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.