Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Acetylated Lysine, Polyclonal

Catalog No. ST102750UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST102750UL 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST102750UL Supplier MilliporeSigma™ Supplier No. ST102750UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Acetylated Lysine Clone: in samples, and it is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Immuno Blotting

Protein acetylation is a reversible modification that can control the activity of histones, p53, and many other proteins. The conserved amino-terminal domains of the four core histones (H2A, H2B, H3 and H4) contain lysines that are acetylated by histone acetyltransferases (HATs) and deacetylated by histone deacetylases (HDACs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetylated Lysine
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In 150mM NaCl, 10mM sodium HEPES, 100μg/mL BSA, 50% glycerol, pH 7.5.
Host Species Rabbit
Immunogen synthetic acetylated-lysine
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes acetylated lysine residues in UV-treated HEK293 cells.
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.