Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylcholine Receptor α Mouse, Unlabeled, Clone: 26, BD

Catalog No. BDB610988 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610988 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610988 Supplier BD Biosciences Supplier No. 610988

Mouse Monoclonal Antibody

Acetylcholine is an amine neurotransmitter at the neuromuscular junction. It is released from the presynaptic membrane of a cholinergic synapse into the synaptic cleft. It diffuses across the cleft and binds acetylcholine receptors (AChR) on the postsynaptic membrane. Receptor binding induces postsynaptic membrane depolarization and the generation of an action potential that produces effects such as muscle contraction. The AChR is a 250kDa pentameric complex of four transmembrane subunits in a stoichiometry of α2βγδ. In response to ligand binding, all subunits participate in the formation of an integral cation channel. However, the acetylcholine binding site is primarily within the α subunit. Myasthenia gravis (MG) is an autoimmune condition in which AchR levels are decreased. Autoantibodies bind and crosslink the AchRs leading to their internalization and degradation. This results in a decreased number of functional AChRs. Patients develop muscular weakness and some voluntary muscle fatigue. However, development of MG is also affected by genetic factors. One of the allelic forms of the AChRα gene appears to significantly contribute to MG susceptibility.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetylcholine Receptor α
Applications Western Blot
Classification Monoclonal
Clone 26
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Acetylcholine Receptor α aa. 332-457
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.