Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylcholinesterase Mouse anti-Bovine, Canine, Chicken, Guinea Pig, Human, Monkey, Clone: AE-1, MilliporeSigma™

Catalog No. MAB303MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB303MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB303MI Supplier MilliporeSigma Supplier No. MAB303
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Acetylcholinesterase Antibody, clone AE-1 is validated for use in ELISA, immunohistochemistry, immunoprecipitation for the detection of Acetylcholinesterase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetylcholinesterase
Applications ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone AE-1
Conjugate Unconjugated
Formulation Liquid in 0.02M PB, 0.25M NaCl, pH 7.6, with 0.1% sodium azide.
Gene Accession No. NM_000665.3; NM_015831.2
Gene Symbols ACHE, N-ACHE, acetylcholinesterase, AChE, YT, ARACHE, EC 3.1.1.7
Host Species Mouse
Immunogen Acetylcholinesterase AChE from human erythrocyte.
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Canine, Chicken, Guinea Pig, Human, Monkey
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.