Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Achaete-scute homolog 1 Mouse anti-Human, Clone: 6D8.1, MilliporeSigma™

Catalog No. MABD64MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD64MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD64MI Supplier MilliporeSigma Supplier No. MABD64

Mouse Monoclonal Antibody

Detect Achaete-scute homolog 1 using this Anti-Achaete-scute homolog 1 Antibody, clone 6D8.1 validated for use in Western Blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Achaete-scute homolog 1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6D8.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_004307
Gene Symbols ASCL1, ASH1, BHLHA46, HASH1
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human Achaete-scute homolog 1.
Purification Method Purified by ion-exchange chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.