Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Achaete Scute homolog 2 Mouse anti-Human, Mouse, Clone: 8F1, MilliporeSigma™

Catalog No. MAB4417MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4417MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4417MI Supplier MilliporeSigma Supplier No. MAB4417
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Achaete Scute homolog 2 Antibody, clone 8F1 is an antibody against Achaete Scute homolog 2 for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Achaete Scute homolog 2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8F1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_032580.2
Gene Symbols ASCL2, ASH2, HASH2, MASH2, Mash2
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to mouse Achaete Scute homolog 2.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.