Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Ceramidase Mouse, Unlabeled, Clone: 23, BD

Catalog No. BDB612302 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612302 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612302 Supplier BD Biosciences Supplier No. 612302
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Ceramide is a sphingolipid that exhibits a wide variety of functions, including monocyte differentiation, apoptosis, neurite outgrowth, and Ca2+ transport. It also serves as the precursor of many sphingolipids and anchors these into the outer leaflet of the plasma membrane via hydrophobic interactions. Acid ceramidase is a lysosomal enzyme that was purified from human urine. It is synthesized as a 55kDa precursor protein, which is then processesed into the mature α-subunit (13kDa) and β-subunit (40kDa). Acid ceramidase catalyzes the hydrolysis of ceramide into free fatty acid and sphingosine. Sphingosine, and its phosphorylated form, sphingosine-1-phosphate (SPP), have been shown to inhibit PKC activity and act as a second messenger in cell proliferation and differentiation. Acid ceramidase is also the cause of a lysosomal storage disorder known as Farber’s disease. This disease is characterized by an accumulation of ceramide in tissues, leading to swelling and pain of the joints and extremities, pulmonary insufficiency, and death at an early age. Thus, acid ceramidase is necessary in the hydrolysis of ceramide and is the cause of Farber’s disease.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Acid Ceramidase
Applications Western Blot
Classification Monoclonal
Clone 23
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Acid Ceramidase aa. 88-182
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.