Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Sensitive Ion Channel 1 Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5674PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5674PMI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5674PMI Supplier MilliporeSigma Supplier No. AB5674P
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Acid Sensitive Ion Channel 1 using this Anti-Acid Sensitive Ion Channel 1 Antibody validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acid Sensitive Ion Channel 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in phosphate buffered saline containing 0.1% BSA.
Gene Symbols ACCN2; BNaC2; ASIC1A; hBNaC2; ASIC1; ASIC; BNAC2
Host Species Rabbit
Immunogen A 20 amino acid peptide sequence near the extracellular N-terminus of rat ASIC1 (Chen et al. 1998; Waldmann et al. 1997; Garcia-Anoveros et al. 1997; Waldmann et al. 1996).

The immunogen sequence has 100% homology with human sodium channel 2 or BNaC2.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001095.2; NM_020039.2
Target Species Human, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.