Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACP2, Mouse, Clone: M1-4A12, Abnova™

Catalog No. 89016024 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-024 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-024 Supplier Abnova Corporation Supplier No. H00000053M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant ACP2.

This gene encodes the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The encoded protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq

Sequence: MAGKRSGWSRAALLQLLLGVNLVVMPPTQARSLRFVTLLYRHGDRSPVKTYPKDPYQEEEWPQGFGQLTKEGMLQHWELGQALRQRYHGFLNTSYHRQEVYVRSTDFDRTLMSAEANLAGLFPPNGMQRFNPNISWQPIPVHTVPITEDRLLKFPLGPCPRYEQLQNETRQTPEYQNESSRNAQFLDMVANETGLTDLTLETVWNVYDTLFCEQTHGLRLPPWASPQTMQRLSRLKDFSFRFLFGIYQQAEKARLQGGVLLAQIRKNLTLMATTSQLPKLLVYSAHDTTLVALQMALDVYNGEQAPYASCHIFELYQEDSGNFSVEMYFRNESDKAPWPLSLPGCPHRCPLQDFLRLTEPVVPKDWQQECQLASGPADTEVIVALAVCGSILFLLIVLLLTVLFRMQAQPPGYRHVADGEDHA

Specifications

Antigen ACP2
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), KnockDown, Western Blot
Classification Monoclonal
Clone M1-4A12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant ACP2.
Formulation PBS with no preservative; pH 7.4
Gene ACP2
Gene Accession No. BC003160
Gene Symbols ACP2
Host Species Mouse
Immunogen ACP2 (AAH03160, 1 a.a. ∼ 423 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.