Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACPP, Mouse, Clone: 2D11, Abnova™

Catalog No. 89016031 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-031 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-031 Supplier Abnova Corporation Supplier No. H00000055M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ACPP.

This gene encodes an enzyme that catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland. An alternatively spliced transcript variant encoding a longer isoform has been found for this gene. This isoform contains a transmembrane domain and is localized in the plasma membrane-endosomal-lysosomal pathway. [provided by RefSeq

Sequence: YASCHLTELYFEKGEYFVEMYYRNETQHEPYPLMLPGCSPSCPLERFAELAGPVIPQDWSTECMTTNSHQVLKVIFAVAFCLISAVLMVLLFIHIRRGLCWQRESYGNI

Specifications

Antigen ACPP
Applications ELISA
Classification Monoclonal
Clone 2D11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ACPP.
Formulation PBS with no preservative; pH 7.4
Gene ACPP
Gene Accession No. BC007460
Gene Alias ACP-3/ACP3/PAP
Gene Symbols ACPP
Host Species Mouse
Immunogen ACPP (AAH07460, 310 a.a. ∼ 418 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.