Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACRV1, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89206329 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-206-329 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-206-329 Supplier Abnova Corporation Supplier No. H00000056K

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human ACRV1 peptide using ARM Technology.

This gene encodes a testis-specific, differentiation antigen, acrosomal vesicle protein 1, that arises within the acrosomal vesicle during spermatogenesis, and is associated with the acrosomal membranes and matrix of mature sperm. This gene consists of 4 exons and its alternative splicing generates multiple distinct transcripts, which encode protein isoforms ranging from 81 to 265 amino acids. The longest transcript is the most abundant, comprising 53-72% of the total acrosomal vesicle protein 1 messages; the second largest transcript comprises 15-32%; the third and the fourth largest transcripts account for 3.4-8.3% and 8.7-12.5%, respectively; and the remaining transcripts combined account for <1% of the total acrosomal vesicle protein 1 message. It is suggested that phenomena of cryptic splicing and exon skipping occur within this gene. The acrosomal vesicle protein 1 may be involved in sperm-zona binding or penetration, and it is a potential contraceptive vaccine immunogen for humans. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen ACRV1
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description acrosomal vesicle protein 1
Formulation In 1X PBS, pH 7.4
Gene ACRV1
Gene Accession No. ACRV1
Gene Alias D11S4365, SP-10, SPACA2
Gene Symbols ACRV1
Host Species Rabbit
Immunogen A synthetic peptide of human ACRV1
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.