Learn More
acyl-CoA synthetase long-chain family member 1, Mouse, Clone: 3G4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ACSL1.
Supplier: Abnova Corporation H00002180M02
Description
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. [provided by RefSeq
Sequence: PKPLKPPCDLSMQSVEVAGSGGARRSALLDSDEPLVYFYDDVTTLYEGFQRGIQVSNNGPCLGSRKPDQPYEWLSYKQVAELSECIGSALIQKGFKTASpecifications
acyl-CoA synthetase long-chain family member 1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001995 | |
ACSL1 | |
ACSL1 (NP_001986, 48 a.a. ∼ 145 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Endocrine/Metabolism | |
2180 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Western Blot | |
3G4 | |
Mouse monoclonal antibody raised against a partial recombinant ACSL1. | |
ACSL1 | |
ACS1/FACL1/FACL2/LACS/LACS1/LACS2 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.