Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Mouse anti-Human, Chicken, AlexaFluor™488, Clone: C4, MilliporeSigma™

Catalog No. MAB1501XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1501XMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1501XMI Supplier MilliporeSigma Supplier No. MAB1501X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Actin Antibody, clone C4, Alexa Fluor 488 conjugated detects level of Actin and has been published and validated for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin
Applications Immunofluorescence
Classification Monoclonal
Clone C4
Conjugate Alexa Fluor 488
Formulation Purified in PBS with 15mg/mL BSA and 0.05% NaN3, conjugated to Alexa Fluor 488.
Gene Accession No. NM_001100.3
Gene Symbols ACTA1, MPFD, ASMA, NEM1, CFTDM, ACTA, CFTD, NEM2, Alpha-actin-1, CFTD1, NEM3
Host Species Mouse
Immunogen Purified chicken gizzard actin (Lessard, 1988).
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Chicken, Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.