Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Mouse anti-Bovine, Chicken, Horse, Human, Mouse, Rat, Clone: ASM-1, MilliporeSigma™

Catalog No. CBL171MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL171MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL171MI Supplier MilliporeSigma Supplier No. CBL171
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Actin Antibody, smooth muscle, clone ASM-1 detects level of Actin and has been published and validated for use in immunofluoresence, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone ASM-1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2a presented as lyophilized material. Reconstitute in 1 mL of of distilled water.
Final solution is PBS, pH 7.4, containing 0.5% BSA and 0.09% sodium azide.
Gene Accession No. NP_001604
Gene Symbols ACTA2, ACTSA, AAT6
Host Species Mouse
Immunogen Synthetic peptide corresponding to the ten N-terminal amino acids of the alpha-smooth muscle isoform of actin.
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species Bovine, Chicken, Equine, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.