Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Actin Mouse anti-All, Clone: C4

Catalog No. MAB1501MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1501MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1501MI Supplier MilliporeSigma™ Supplier No. MAB1501
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reliably and specifically detect actin using this Anti-Actin Antibody, clone C4. This highly published monoclonal antibody is validated for use in ELISA, immunocytochemistry, immunofluoresence, immunohistochemistry, immunohistochemistry (paraffin) and western blotting. This mAb is also available as a fluorescent conjugate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C4
Conjugate Unconjugated
Formulation Mouse monoclonal Ascites fluid, with 0.01% sodium azide.
Gene Accession No. NP_001091.1
Gene Symbols ACTA1, MPFD, ASMA, NEM1, CFTDM, ACTA, CFTD, NEM2, Alpha-actin-1, CFTD1, NEM3
Host Species Mouse
Immunogen Purified chicken gizzard actin (Lessard, 1988).
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species All species
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.