Learn More
MilliporeSigma™ anti-Actin, Clone: C4, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1501/DEL

Description
Actin is an abundant cytoskeletal protein found in all cells (True, 1990). The protein's 42 kD peptide chain assumes two physical forms: globular actin, which may serve as a cytoplasmic storage pool, and fibrous actin, which, in conjunction with myosin, generates muscle contraction (True, 1990). In non-muscle cells, actin appears to be involved in a variety of functions, such as cell motility, exocytosis, and phagocytosis (True, 1990). Distribution of the six known isoforms of actin - four muscle actins (α-skeletal, α-vascular smooth, α-cardiac, and γ-enteric smooth) and two cytoplasmic actins (α and γ) is tissue specific (Otey, 1986; Lessard, 1988).
Lessard, 1988.
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting

Specifications
Actin | |
Unconjugated | |
Purified chicken gizzard actin | |
RUO | |
Primary | |
IgG1 κ |
Monoclonal | |
Mouse | |
100 μL | |
Cell Structure | |
Ascites |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.