Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Mouse anti-Chicken, Human, Monkey, Rat, Rabbit, Clone: CGA7, MilliporeSigma™

Catalog No. MAB1522MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1522MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1522MI Supplier MilliporeSigma Supplier No. MAB1522
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Actin using this Anti-Actin Antibody, smooth muscle γ and α actin, clone CGA7 validated for use in western blot, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CGA7
Conjugate Unconjugated
Formulation Ascites. Liquid
Gene Accession No. NM_001613.1
Gene Symbols ACTA2, Alpha-actin-2, ACTSA
Host Species Mouse
Immunogen Chicken gizzard actin
Quantity 100 μL
Primary or Secondary Primary
Target Species Chicken, Human, Monkey, Rabbit, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.