Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Active-Β-Catenin (ABC) Mouse anti-Human, Mouse, Rat, Clone: 8E7

Catalog No. 05665MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-665-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-665-MI Supplier MilliporeSigma™ Supplier No. 05665
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Active-β-Catenin (anti-ABC) Antibody, clone 8E7 is a well characterized Mouse Monoclonal Antibody. This highly published mAb also known as Anti-Catenin beta-1 readily detects beta-Catenin and has been validated in flow cytometry, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Active-β-Catenin (ABC)
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8E7
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1k in buffer containing 0.1MTris-Glycine, pH 7.4, 15mM NaCl and 0.05% sodium azide
Gene Accession No. NP_001895
Gene Symbols CTNNB1, CTNNB, FLJ25606, Beta-catenin, PRO2286, FLJ37923, DKFZp686D02253
Host Species Mouse
Immunogen Fusion protein corresponding to residues 1-100 of human β-catenin with a c-terminal histidine tag
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.