Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-ACTN4 Clone: 4D10,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ CB1024100UG
Description
Specifically detects ACTN4 Clone: 4D10 in Human, Mouse, Rat samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immuno Blotting
Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In non-muscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments.
Specifications
ACTN4 | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
ELISA, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
4D10 | |
In PBS, pH 7.2. | |
A recombinant polypeptide corresponding to amino acids 592-702 of human ACTN4, espressed as a GST fusion protein | |
RUO | |
Recognizes the ∽90-100kDa ACTN4 protein in PC-12, HeLa, Raw 264.7, MCF-7, and NIH3T3 cells and human testes tissue. | |
−20°C | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction