Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACYP1, Mouse, Clone: 1B2-3A2, Abnova™

Catalog No. 89016047 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-047 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-047 Supplier Abnova Corporation Supplier No. H00000097M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant ACYP1.

Acylphosphatase is a small cytosolic enzyme that catalyzes the hydrolysis of the carboxyl-phosphate bond of acylphosphates. Two isoenzymes have been isolated, called muscle acylphosphatase and erythrocyte acylphosphatase, on the basis of their tissue localization. This gene encodes the erythrocyte acylphosphatase isoenzyme. Alternatively spliced transcript variants that encode different proteins were identified through data analysis. [provided by RefSeq

Sequence: MAEGNTLISVDYEIFGKVQGVFFRKHTQAEGKKLGLVGWVQNTDRGTVQGQLQGPISKVRHMQEWLETRGSPKSHIDKANFNNEKVILKLDYSDFQIVK

Specifications

Antigen ACYP1
Applications ELISA, Western Blot
Classification Monoclonal
Clone S3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant ACYP1.
Formulation PBS with no preservative; pH 7.4
Gene ACYP1
Gene Accession No. BC035568
Gene Alias ACYPE
Gene Symbols ACYP1
Host Species Mouse
Immunogen ACYP1 (AAH35568, 1 a.a. ∼ 99 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 97
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.