Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM 10 Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB19026MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB19026MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB19026MI Supplier MilliporeSigma Supplier No. AB19026
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-ADAM 10 Antibody, C-terminus is validated for use in western blotting for the detection of ADAM 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM 10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing PBS and 0.02% sodium azide.
Gene Symbols AD10; CD156c; CDw156; EC 3.4.24.81; HsT18717; KUZ; MADM; kuz; kuzbanian
Host Species Rabbit
Immunogen Raised against a peptide corresponding to amino acids 732-748 of human ADAM 10 (Rosendahl, M.S., 1997). This sequence is identical to those of bovine and rat origins and differs from that of mouse ADAM 10 by one amino acid (Pan, D., 1997; Howard, L., 199
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001110.2
Target Species Bovine, Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.