Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM 12 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB19032
Description
Detect ADAM 12 using this Anti-ADAM 12 Antibody, C-terminus validated for use in western blotting.
Specifications
ADAM 12 | |
Polyclonal | |
Neat rabbit serum. Contains no preservative. | |
Rabbit | |
100 μL | |
NM_003474.3; NM_021641.2 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Western Blot | |
Unconjugated | |
ADAM12; Meltrin-alpha; MLTN; MCMP; MLTNA; Mltna; OTTHUMP00000046766; MCMPMltna; EC 3.4.24.- | |
13 amino acid synthetic peptide from the C-terminous of the protein (HQVPRPSHNAYIK) | |
Primary | |
Mouse | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction