Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adam17 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABT94MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT94MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT94MI Supplier MilliporeSigma Supplier No. ABT94
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Adam17 Antibody is a highly specific rabbit polyclonal antibody, that targets ADAM 17 and has been tested in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adam17
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Symbols Adam17; TACE
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the cytoplasmic domain of mouse Adam17.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_033745
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.