Learn More
ADAM2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ADAM2 peptide using ARM Technology.
Supplier: Abnova Corporation H00002515K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
ADAM2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
ADAM2 | |
ADAM2 | |
A synthetic peptide of human ADAM2 | |
RUO | |
2515 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
ADAM metallopeptidase domain 2 | |
ADAM2 | |
CRYN1, CRYN2, FTNB, PH-30b, PH30 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.