Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM9 Mouse, Unlabeled, Clone: 15, BD

Catalog No. BDB610750 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610750 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610750 Supplier BD Biosciences Supplier No. 610750

Mouse Monoclonal Antibody

MDC9 (metalloprotease/disintegrin/cysteine-rich protein 9) belongs to a family of cellular disintegrins known as ADAM (A Disintegrin And Metalloprotease domain). The ADAM proteins are involved in cell-cell and cell-matrix adhesion and, possibly, the degradation of the extracellular matrix. All members possess: 1) a prodomain; 2) a metalloprotease-, disintegrin-, and EGF-like region; 3) a cysteine-rich region; 4) a transmembrane domain; and 5) a cytoplasmic domain. In addition to these regions, MDC9 contains two cytoplasmic proline-rich sequences that may bind to the SH3 domains of cytoskeletal or signaling proteins. MDC9 also contains a membrane-bound metalloprotease and disintegrin domain, suggesting multifunctional activity. Immunofluorescence staining demonstrated that MDC9 is located at the plasma membrane. MDC9 human and mouse protein sequence share 82% identity and are ubiquitously expressed.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM9
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 15
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human MDC9 aa. 86-227
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.