Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS-1 Mouse anti-Human, Mouse, Rat, Clone: 5D4E11B6, MilliporeSigma™

Catalog No. MAB1810MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1810MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1810MI Supplier MilliporeSigma Supplier No. MAB1810
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect ADAMTS-1 using this Anti-ADAMTS-1 Antibody, clone 5D4E11B6 validated for use in ELISA, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS-1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5D4E11B6
Conjugate Unconjugated
Formulation Ascites. Liquid containing 0.05% NaN3.
Gene Accession No. NM_006988.3
Gene Symbols ADAMTS1, KIAA1346, METH1, C3-C5
Host Species Mouse
Immunogen Myc-His carboxyl tagged full-length recombinant protein based on the human sequence.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.