Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM metallopeptidase with thrombospondin type 1 motif, 18, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016631 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-631 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-631 Supplier Abnova Corporation Supplier No. H00170692B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human ADAMTS18 protein.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has a high sequence similarity to the protein encoded by gene ADAMTS16, another family member. It is thought to function as a tumor suppressor. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined. [provided by RefSeq

Sequence: MECALLLACAFPAAGSGPPRGLAGLGRVAKALQLCCLCCASVAAALASDSSSGASGLNDDYVFVTPVEVDSAGSYISHDILHNGRKKRSAQNARSSLHYRFSAFGQELHLELKPSAILSSHFIVQVLGKDGASETQKPEVQQCFYQGFIRNDSSSSVAVSTCAGLSGLIRTRKNEFLISPLPQLLAQEHNHSSPAGHHPHVLYKRTAEEKIQRYRGYPGSGRNYPGYSPSHIPHASQSRETEYHHRRLQKQHFCGRRKKYAPKPPTEDTYLRFDEYGSSGRPRRSAGKSQKGLNVETLVVADKKMVEKHGKGNVTTYILTVMNMVSGLFKDGTIGSDINVVVVSLILLEQEPGGLLINHHADQSLNSFCQWQSALIGKNGKRHDHAILLTGFDICSWKNEPCDTLGFAPISGMCSKYRSCTINEDTGLGLAFTIAHESGHNFGMIHDGEGNPCRKAEGNIMSPTLTGNNGVFSWSSCSRQYLKKFLSTPQAGCLVDEPKQAGQYKYPDKLPGQIYDADTQCKWQFGAKAKLCSLGFVKDICKSLWCHRVGHRCETKFMPAAEGTVCGLSMWCRQGQCVKFGELGPRPIHGQWSAWSKWSECSRTCGGGVKFQERHCNNPKPQYGGIFCPGSSRIYQLCNINPCNENSLDFRAQQCAEYNSKPFRGWFYQWKPYTKVEEEDRCKLYCKAENFEFFFAMSGKVKDGTPCSPNKNDVCIDGVCELVGCDHELGSKAVSDACGVCKGDNSTCKFYKGLYLNQHKANEYYPVVIIPAGARSIEIQELQVSSSYLAVRSLSQKYYLTGGWSIDWPGEFPFAGTTFEYQRSFNRPERLYAPGPTNETLVFEILMQGKNPGIAWKYALPKVMNGTPPATKRPAYTWSIVQSECSVSCGGGYINVKAICLRDQNTQVNSSFCSAKTKPVTEPKICNAFSCPAYWMPGEWSTCSKACAGGQQSRKIQCVQKKPFQKEEAVLHSLCPVSTPTQVQACNSHACPPQWSLGPWSQCSKTCGRGVRKRELLCKGSAAETLPESQCTSLPRPELQEGCVLGRCPKNSRLQWVASSWSECSATCGLGVRKREMKCSEKGFQGKLITFPERRCRNIKKPNLDLEETCNRRACPAHPVYNMVAGWYSLPWQQCTVTCGGGVQTRSVHCVQQGRPSSSCLLHQKPPVLRACNTNFCPAPEKREDPSCVDFFNWCHLVPQHGVCNHKFYGKQCCKSCTRKI

Specifications

Antigen ADAM metallopeptidase with thrombospondin type 1 motif, 18
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ADAMTS18 protein.
Formulation PBS with no preservative; pH 7.4
Gene ADAMTS18
Gene Accession No. BC063283.1
Gene Alias ADAMTS21
Gene Symbols ADAMTS18
Host Species Mouse
Immunogen ADAMTS18 (AAH63283.1, 1 a.a. ∼ 1221 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170692
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.