Learn More
Adaptin γ Mouse, Unlabeled, Clone: 88, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610386
Description
Sorting of integral membrane proteins at various stages of the endocytic and secretory pathways is mediated by vesicular trafficking between a variety of organelles. Two sorting signals are tyrosine-based and dileucine-based signals that interact with heterotetrameric adaptor protein complexes (AP-1, AP-2, AP-3, and AP-4), which are associated with the vesicle coats. These coatomers contain two large Adaptin proteins (γ, α, δ, or ε and β1, β2, β3, β4 respectively) that are noncovalently linked to one medium chain (μ1, μ2, μ3, or μ4) and one small chain ( σ1, σ2, σ3, or σ4). The AP-1 and AP-3 complexes are involved in protein sorting from the TGN and endosomes, while AP-2 adaptor complexes are involved in clathrin-mediated endocytosis. Adaptin ? shows more homology with Adaptin α than with Adaptin β. The conserved regions between Adaptins γ and α could be important for binding to other components of the AP-1 and AP-2 complexes, respectively.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| Adaptin γ | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Mouse Adaptin γ aa. 642-821 | |
| 150 μg | |
| Cell Biology | |
| Canine, Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 88 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.