Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAR2 Mouse anti-Human, Clone: 1.3.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE889
Description
Anti-ADAR2 Antibody, clone 1.3.1 detects level of ADAR2 and has been published and validated for use in ADAR2.
Specifications
| ADAR2 | |
| Monoclonal | |
| Unconjugated | |
| NP_001103 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| 2-8°C for one year | |
| IgG1 κ |
| Western Blot | |
| 1.3.1 | |
| Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| ADARB1, ADAR2, DRADA2, RED1 | |
| GST-tagged recombinant protein corresponding to the C-terminus of human ADAR2. | |
| 100 μg | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction