Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAR2 Mouse anti-Human, Clone: 2D1.1, MilliporeSigma™

Catalog No. MABE355MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE355MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE355MI Supplier MilliporeSigma Supplier No. MABE355
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-ADAR2 Antibody, clone 2D1.1 is an antibody against ADAR2 for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAR2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2D1.1
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_005378
Gene Symbols NUP98, ADAR2
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human ADAR2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.