Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCY2, Mouse, Clone: 1D4, Abnova™

Catalog No. 89016049 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-049 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-049 Supplier Abnova Corporation Supplier No. H00000108M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ADCY2.

This gene encodes a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This enzyme is insensitive to Ca(2+)/calmodulin, and is stimulated by the G protein beta and gamma subunit complex. [provided by RefSeq]

Sequence: GKLDAINKHSFNDFKLRVGINHGPVIAGVIGAQKPQYDIWGNTVNVASRMDSTGVLDKIQVTEETSLVLQTLGYTCTCRGIINVKGKGDLKTYFVNTEMSRSLSQSNVAS

Specifications

Antigen ADCY2
Applications ELISA, In situ PLA
Classification Monoclonal
Clone 1D4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ADCY2.
Formulation PBS with no preservative; pH 7.4
Gene ADCY2
Gene Accession No. NM_020546
Gene Alias AC2/FLJ16822/FLJ45092/HBAC2/KIAA1060/MGC133314
Gene Symbols ADCY2
Host Species Mouse
Immunogen ADCY2 (NP_065433, 977 a.a. ∼ 1086 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.