Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADCYAP1R1, Mouse, Clone: 2B12, Abnova™

Catalog No. 89016050 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-050 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-050 Supplier Abnova Corporation Supplier No. H00000117M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ADCYAP1R1.

This gene encodes type I adenylate cyclase activating polypeptide receptor, which is a membrane-associated protein and shares significant homology with members of the glucagon/secretin receptor family. This receptor mediates diverse biological actions of adenylate cyclase activating polypeptide 1 and is positively coupled to adenylate cyclase. Alternative splicing of two exons of this gene generates four major splice variants, but their full-length nature has not been determined. [provided by RefSeq

Sequence: MHSDCIFKKEQAMCLEKIQRANELMGFNDSSPGCPGMWDNITCWKPAHVGEMVLVSCPELFRIFNPDQVWETETIGESDFGDSNSLDLSDMGVVSRNCTE

Specifications

Antigen ADCYAP1R1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ADCYAP1R1.
Formulation PBS with no preservative; pH 7.4
Gene ADCYAP1R1
Gene Accession No. NM_001118
Gene Alias PAC1/PACAPR/PACAPRI
Gene Symbols ADCYAP1R1
Host Species Mouse
Immunogen ADCYAP1R1 (NP_001109, 21 a.a. ∼ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.