Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A2a Receptor Rabbit anti-Bovine, Canine, Human, Pig, Rat, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

Supplier:  MilliporeSigma AB1559

Catalog No. AB1559MI


Only null left
Add to Cart

Description

Description

This Anti-Adenosine A2a Receptor Antibody is validated for use in ELISA, western blotting for the detection of Adenosine A2a Receptor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adenosine A2a Receptor
Polyclonal
Rabbit antiserum. Liquid containing 0.05% sodium azide.
Rabbit
50 μL
NM_000675.4
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
ELISA, Western Blot
Unconjugated
ADORA2A; ADORA2; RDC8; hA2aR
Synthetic 30 amino acid peptide corresponding to amino acids 383-412 of the canine adenosine A2a receptor near the C-terminal intracellular domain (30 amino acids) of the cloned canine A2a receptor. The A2a sequence selected in not present in A1, A2b and
Primary
Bovine, Canine, Human, Pig, Rat
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only