Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine A2b Receptor Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1589P
Description
Detect Adenosine A2b Receptor using this Anti-Adenosine A2b Receptor Antibody validated for use in ELISA, immunocytochemistry, immunohistochemistry and western blotting.
Specifications
Adenosine A2b Receptor | |
Polyclonal | |
Affinity purified immunoglobulin. Liquid in PBS with 0.1% BSA. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
ADORA2B; ADORA2 | |
Amino acid sequence corresponding to the 2nd extracellular domain (16 amino acids) of the human brain A2bR gene (Pierce et al. 1992; Tucker & Linden 1993). | |
50 μg | |
NM_000676.2 | |
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction