Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine Receptor A2a Mouse anti-Human, Rat, Mouse, Rabbit, Canine, Clone: 7F6-G5-A2, MilliporeSigma™

Catalog No. 05717MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-717-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-717-MI Supplier MilliporeSigma Supplier No. 05717
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Adenosine Receptor A2a Antibody, clone 7F6-G5-A2 is an antibody against Adenosine Receptor A2a clone 7F6-G5-A2 for use in flow cytometry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenosine Receptor A2a
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 7F6-G5-A2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2ak in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide
Gene Accession No. NM_000675
Gene Symbols ADORA2A, ADORA2, RDC8, hA2aR
Host Species Mouse
Immunogen 6His-tagged full-length human recombinant A2A receptor. Epitope mapped to SQPLPGER in the third intracellular loop. Clone 7F6-G5-A2.
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rabbit, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.