Learn More
adipose differentiation-related protein, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
Adipocyte differentiation-related protein is associated with the globule surface membrane material. This protein is a major constituent of the globule surface. Increase in mRNA levels is one of the earliest indications of adipocyte differentiation [provided by RefSeq
Specifications
Specifications
| Antigen | adipose differentiation-related protein |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length human ADFP protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ADFP |
| Gene Accession No. | NM_001122.2 |
| Gene Alias | ADRP/MGC10598 |
| Gene Symbols | ADFP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.