Learn More
ADH4, Mouse, Clone: 1D2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ADH4.
Supplier: Abnova Corporation H00000127M03A
Description
This gene encodes class II alcohol dehydrogenase 4 pi subunit, which is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Class II alcohol dehydrogenase is a homodimer composed of 2 pi subunits. It exhibits a high activity for oxidation of long-chain aliphatic alcohols and aromatic alcohols and is less sensitive to pyrazole. This gene is localized to chromosome 4 in the cluster of alcohol dehydrogenase genes. [provided by RefSeq
Sequence: SVIDSKFEGLAFPVIVGHEAAGIVESIGPGVTNVKPGDKVIPLYAPLCRKCKFCLSPLTNLCGKISNLKSPASDQQLMEDKTSRFTCKGKPVYHFFGTSSpecifications
ADH4 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_000670 | |
ADH4 | |
ADH4 (NP_000661, 52 a.a. ∼ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
127 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
1D2 | |
Mouse monoclonal antibody raised against a partial recombinant ADH4. | |
ADH4 | |
ADH-2 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.