Learn More
ADH6, Mouse, Clone: 4G4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ADH6.
Supplier: Abnova Corporation H00000130M01
Description
This gene encodes class V alcohol dehydrogenase, which is a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. This gene is expressed in the stomach as well as in the liver, and it contains a glucocorticoid response element upstream of its 5' UTR, which is a steroid hormone receptor binding site. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: GSKHLDLLYPTILGHEGAGIVESIGEGVSTVKPGDKVITLFLPQCGECTSCLNSEGNFCIQFKQSKTQLMSDGTSRFTCKGKSIYHFGNTSpecifications
ADH6 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000672 | |
ADH6 | |
ADH6 (NP_000663, 55 a.a. ∼ 144 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4G4 | |
Mouse monoclonal antibody raised against a partial recombinant ADH6. | |
ADH6 | |
ADH-5 | |
Mouse | |
Affinity chromatography | |
RUO | |
130 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.