Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADK, Mouse, Clone: 4E7, Abnova™

Mouse monoclonal antibody raised against a partial recombinant ADK.

Supplier:  Abnova Corporation H00000132M01

Catalog No. 89-016-056


Only null left
Add to Cart

Description

Description

This gene encodes adenosine kinase, an abundant enzyme in mammalian tissues. The enzyme catalyzes the transfer of the gamma-phosphate from ATP to adenosine, thereby serving as a regulator of concentrations of both extracellular adenosine and intracellular adenine nucleotides. Adenosine has widespread effects on the cardiovascular, nervous, respiratory, and immune systems and inhibitors of the enzyme could play an important pharmacological role in increasing intravascular adenosine concentrations and acting as anti-inflammatory agents. Alternative splicing results in two transcript variants encoding different isoforms. Both isoforms of the enzyme phosphorylate adenosine with identical kinetics and both require Mg2+ for activity. [provided by RefSeq]

Sequence: FETKDIKEIAKKTQALPKMNSKRQRIVIFTQGRDDTIMATESEVTAFAVLDQDQKEIIDTNGAGDAFVGGFLSQLVSDKPLTECIRAGHYAASIIIRRTGCTFPEKPDFH
Specifications

Specifications

ADK
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_001123
ADK
ADK (NP_001114, 236 a.a. ∼ 345 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA
4E7
Mouse monoclonal antibody raised against a partial recombinant ADK.
ADK
AK
Mouse
Affinity chromatography
RUO
132
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.