Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADORA2A, Mouse, Clone: 4E4, Abnova™

Catalog No. 89123136 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-136 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-136 Supplier Abnova Corporation Supplier No. H00000135M17
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ADORA2A.

This gene encodes a protein which is one of several receptor subtypes for adenosine. The activity of the encoded protein, a G-protein coupled receptor family member, is mediated by G proteins which activate adenylyl cyclase. The encoded protein is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine. [provided by RefSeq

Sequence: GQPKEGKNHSQGCGEGQVACLFEDVVPMNYMVYFNFFACVLVPLLLMLGVYLRIFLAARRQLKQMESQPLPGERARSTLQKEVHAAKSLAIIVGLFALCWLPLHIINCFTFFCPDCSHAPL

Specifications

Antigen ADORA2A
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4E4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ADORA2A.
Formulation PBS with no preservative; pH 7.4
Gene ADORA2A
Gene Accession No. NM_000675.3
Gene Alias ADORA2/RDC8/hA2aR
Gene Symbols ADORA2A
Host Species Mouse
Immunogen ADORA2A (NP_000666.2, 147 a.a. ∼ 267 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.